n-Undecane ≥99%
Supplier: BeanTown Chemical
Synonyms:
Undecane
123160-250G
123160-50G
BT123160-250GEA
139
USD
BT123160-250G
BT123160-50G
n-Undecane ≥99%
n-Undecane
CAS: 1120-21-4; EC No: 214-300-6; MDL No: MFCD00008959; RTECS: YQ1525000
UN No: UN2330; Haz Class: 3; Packing Group: III
Liquid; Linear Formula: CH3(CH2)9CH3; Molecular Formula: C11H24; MW: 156.31
Melting Point: -26°; Boiling Point: 196°; Flash point: 60°C (140°F)
Density (g/mL): 0.74; Refractive Index: 1.417
- RTECS: YQ1525000
- Refractive index: 1.417
- Liquid
Formula:
H₃C(CH₂)₉CH₃ MW: 156.31 g/mol Boiling Pt: 196 Melting Pt: -26.7 Density: 0.74 g/mL Flash Pt: 60 °C (140 °F) |
MDL Number:
MFCD00008959 CAS Number: 1120-21-4 EINECS: 214-300-6 UN: 2330 ADR: 3 III |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...