Triethylamine ≥99%

Supplier: BeanTown Chemical

Synonyms: (Diethylamino)ethane, TEA

122970-1KG 122970-250G
BT122970-1KGEA 75.99 USD
BT122970-1KG BT122970-250G
Triethylamine ≥99%
Triethylamine

CAS: 121-44-8; EC No: 204-469-4; MDL No: MFCD00009051; RTECS: YE0175000 UN No: UN1296; Haz Class: 3 (8); Packing Group: II Liquid; Linear Formula: (CH3CH2)3N; Molecular Formula: C6H15N; MW: 101.19 Melting Point: -115°; Boiling Point: 89°; Flash point: -15°C (5°F) Density (g/mL): 0.726; Refractive Index: 1.401


  • RTECS: YE0175000
  • Refractive index: 1.401
  • Liquid

Formula: (C₂H₅)₃N
MW: 101.19 g/mol
Boiling Pt: 90
Melting Pt: -115
Density: 0.726 g/mL
Flash Pt: –15 °C (5 °F)
MDL Number: MFCD00009051
CAS Number: 121-44-8
EINECS: 204-469-4
UN: 1296
ADR: 3 II
Merck Index: 13,09740
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c2b1f184e438ecb01f88a4d90ee46c0d4316859d&vid=1c68408a0eedf1bfdb02d1b1e2337d50d449cf9d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR