Silver sulfate ≥98.0% ACS

Supplier: BeanTown Chemical

Synonyms: Disilver sulfate, Silver(I) sulfate, Disilver sulphate

122485-100G 122485-25G
BT122485-100GEA 352.64 USD
BT122485-100G BT122485-25G
Silver sulfate ≥98.0% ACS
Silver sulphate

CAS: 10294-26-5; EC No: 233-653-7; MDL No: MFCD00003407 UN No: UN3077; Haz Class: 9; Packing Group: III Crystalline/Powder; Linear Formula: Ag2SO4; MW: 311.80 Melting Point: 652°; Boiling Point: 1085° (decomposes) Density (g/mL): 5.45 Light Sensitive


  • Light Sensitive
  • Crystalline/Powder

Formula: Ag₂SO₄
MW: 311.8 g/mol
Boiling Pt: 1085 (dec.)
Melting Pt: 655
Density: 5.45 g/mL
MDL Number: MFCD00003407
CAS Number: 10294-26-5
EINECS: 233-653-7
ADR: 9 III
Merck Index: 13,08603
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dd7afaa8e9ae24bf040d40aed0c93763ce897080&vid=ae7b50634b53507047974c2157e5e38c2eb90a20&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 98.0% min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR