Diphenyl ether ≥99%
Supplier: BeanTown Chemical
Synonyms:
Diphenyl oxide, Phenyl ether, Phenoxybenzene
122425-2.5KG
122425-500G
BT122425-2.5KGEA
94
USD
BT122425-2.5KG
BT122425-500G
Diphenyl ether ≥99%
Diphenyl ether
CAS: 101-84-8; EC No: 202-981-2; MDL No: MFCD00003034; RTECS: KN8970000
UN No: UN3077; Haz Class: 9; Packing Group: III
Liquid/Low Melting Solid; Linear Formula: (C6H5)2O; MW: 170.21
Melting Point: 25-27°; Boiling Point: 259°; Flash point: 115°C (239°F)
Density (g/mL): 1.073; Refractive Index: 1.579
- RTECS: KN8970000
- Refractive index: 1.579
- Liquid/Low Melting Solid
Formula:
C₁₂H₁₀O Boiling Pt: 259 °C Melting Pt: 25…27 °C Density: 1.073 g/mL Flash Pt: 115 °C (239 °F) |
MDL Number:
MFCD00003034 CAS Number: 101-84-8 EINECS: 202-981-2 UN: 3077 ADR: 9,III |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...