Colchicine ≥95%

Supplier: BeanTown Chemical

Synonyms: 7α-H-Colchicine, (S)-(-)-Colchicine

122410-1G 122410-5G
BT122410-1GEA 91 USD
BT122410-1G BT122410-5G
Colchicine ≥95%
Colchicine

CAS: 64-86-8; EC No: 200-598-5; MDL No: MFCD00078484; RTECS: GH0700000 UN No: UN1544; Haz Class: 6.1; Packing Group: I Powder; Molecular Formula: C22H25NO6; MW: 399.44 Melting Point: 150-160° (decomposes) Light Sensitive


  • RTECS: GH0700000
  • Light Sensitive
  • Powder

Formula: C₂₂H₂₅NO₆
MW: 399.44 g/mol
Melting Pt: 150…160 °C (dec.)
MDL Number: MFCD00078484
CAS Number: 64-86-8
EINECS: 200-598-5
UN: 1544
ADR: 6.1 I
Merck Index: 12,02536

Order Now

Specification Test Results

Assay 95%

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR