Copper, soluble -325 mesh

Supplier: BeanTown Chemical

122105-100G 122105-1KG 122105-500G
BT122105-100GEA 49 USD
BT122105-100G BT122105-1KG BT122105-500G
Copper, soluble -325 mesh
Copper

CAS: 7440-50-8; EC No: 231-159-6; MDL No: MFCD00010965 UN No: UN3089; Haz Class: 4.1; Packing Group: III Solid, average thickness 1-2 microns; Molecular Formula: Cu; MW: 63.55 Melting Point: 1083.4°; Boiling Point: 2567° Density (g/mL): 8.94


  • Solid, average thickness 1-2 microns

Formula: Cu
MW: 63.55 g/mol
Boiling Pt: 2595
Melting Pt: 1083
Density: 8.94 g/mL
MDL Number: MFCD00010965
CAS Number: 7440-50-8
EINECS: 231-159-6
ADR: 4.1 III
Merck Index: 13,02545

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR