Methanesulfonic acid ≥98%

Supplier: BeanTown Chemical

121975-100G 121975-2.5KG 121975-500G
BT121975-100GEA 32.74 USD
BT121975-100G BT121975-2.5KG BT121975-500G
Methanesulfonic acid ≥98%
Methanesulphonic acid

CAS: 75-75-2; EC No: 200-898-6; MDL No: MFCD00007518; RTECS: PB1140000 UN No: UN2586; Haz Class: 8; Packing Group: III Liquid; Linear Formula: CH3SO3H; MW: 96.10 Melting Point: 17-19°; Boiling Point: 166-168°/10 mmHg; Flash point: 156°C (312°F) Density (g/mL): 1.481; Refractive Index: 1.429 Heat Sensitive, Hygroscopic, Light Sensitive


  • RTECS: PB1140000
  • Refractive index: 1.429
  • Heat Sensitive, Hygroscopic, Light Sensitive
  • Liquid

Formula: CH₃SO₃H
Boiling Pt: 166…168 °C/10 mmHg
Melting Pt: 17…19 °C
Density: 1.481 g/mL
Flash Pt: 156 °C (312 °F)
MDL Number: MFCD00007518
CAS Number: 75-75-2
EINECS: 200-898-6
UN: 2586
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bebd3f86cbdbb894828247119a403c686b0ecddf&vid=01bb7db9690bd0b27a8ab4f277f8a301da72087f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR