4,4'-Bi-o-toluidine dihydrochloride ACS

Supplier: BeanTown Chemical

Synonyms: o-Toluidine dihydrochloride, o-Tolidine dihydrochloride

121900-100G 121900-25G 121900-500G
BT121900-100GEA 124 USD
BT121900-100G BT121900-25G BT121900-500G
4,4'-Bi-o-toluidine dihydrochloride ACS
4,4'-Bi-o-toluidine dihydrochloride

CAS: 612-82-8; EC No: 210-322-5; MDL No: MFCD00012960; RTECS: DD1226000 UN No: UN3077; Haz Class: 9; Packing Group: III Solid; Molecular Formula: C14H16N2·2HCl ; MW: 285.22 Melting Point: 300° Air Sensitive


  • RTECS: DD1226000
  • Air Sensitive
  • Solid

Formula: C₁₄H₁₈Cl₂N₂
MW: 285.22 g/mol
Melting Pt: 130
MDL Number: MFCD00012960
CAS Number: 612-82-8
EINECS: 210-322-5
ADR: 9 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR