Acetyl chloride ≥99%

Supplier: BeanTown Chemical

Synonyms: Acetyl chloride 1M in Dichloromethane

121720-1L 121720-250ML 121720-4L 121720-50ML
BT121720-1LEA 169 USD
BT121720-1L BT121720-250ML BT121720-4L BT121720-50ML
Acetyl chloride ≥99%
Acetyl chloride

CAS: 75-36-5; EC No: 200-865-6; MDL No: MFCD00000719; RTECS: AO6390000 UN No: UN1717; Haz Class: 3 (8); Packing Group: II Liquid; Linear Formula: CH3COCl ; MW: 78.50 Melting Point: -112°; Boiling Point: 52°; Flash point: 5°C (41°F) Density (g/mL): 1.104; Refractive Index: 1.389 Moisture Sensitive


  • RTECS: AO6390000
  • Refractive index: 1.389
  • Moisture Sensitive
  • Liquid

Formula: C₂H₃ClO
MW: 78.5 g/mol
Boiling Pt: 50.9
Melting Pt: -112
Density: 1.104 g/mL
Flash Pt: 5 °C (41 °F)
MDL Number: MFCD00000719
CAS Number: 75-36-5
EINECS: 200-865-6
UN: 1717
ADR: 3 II
Merck Index: 13,00087

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR