Methanol ≥99.9% semiconductor grade
Supplier: BeanTown Chemical
121365-100ML
121365-1L
121365-4L
BT121365-100MLEA
31
USD
BT121365-100ML
BT121365-1L
BT121365-4L
Methanol ≥99.9% semiconductor grade
Methanol
CAS: 67-56-1; EC No: 200-659-6; MDL No: MFCD00004595; RTECS: PC1400000
UN No: UN1230; Haz Class: 3; Packing Group: II
Liquid; Linear Formula: CH3OH; MW: 32.04
Melting Point: -98°; Boiling Point: 64.7°; Flash point: 9.7°C (49.5°F)
Density (g/mL): 0.791; Refractive Index: 1.329
- RTECS: PC1400000
- Refractive index: 1.329
- Liquid
Formula:
CH₃OH Boiling Pt: 64.7 °C Melting Pt: –98 °C Density: 0.791 g/mL Flash Pt: 9.7 °C (49.5 °F) |
MDL Number:
MFCD00004595 CAS Number: 67-56-1 EINECS: 200-659-6 UN: 1230 ADR: 3,II |
|
Specification Test Results
Assay | 99.9% min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...