Magnesium nitrate hydrate ≥99.999%

Supplier: BeanTown Chemical
Warning

121060-100G 121060-25G 121060-5G
BT121060-100GEA 249 USD
BT121060-100G BT121060-25G BT121060-5G
Magnesium nitrate hydrate ≥99.999%
Magnesium nitrate hydrate

CAS: 10377-60-3; EC No: 233-826-7; MDL No: MFCD00011103; RTECS: OM3756000 UN No: UN1474; Haz Class: 5.1; Packing Group: III Crystalline Aggregates; Linear Formula: Mg(NO3)2·xH2O; MW: 148.32 (anhydrous) Melting Point: 89°; Boiling Point: 330° (decomposes) Density (g/mL): 1.46 Hygroscopic


  • RTECS: OM3756000
  • Crystalline Aggregates
  • Hygroscopic

Formula: Mg(NO₃)₂·xH₂O
MW: 148.32 g/mol (anhy)
Boiling Pt: 330 °C (dec.)
Melting Pt: 89 °C
Density: 1.46 g/mL
MDL Number: MFCD00011103
CAS Number: 10377-60-3
EINECS: 233-826-7
UN: 1474
ADR: 5.1 III

Order Now

Specification Test Results

Assay 99.999%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR