Zinc chloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Butter of zincZinc (II) chloride

Z0014-300G Z0014-25G
TCZ0014-300GEA 64.27 USD
TCZ0014-300G TCZ0014-25G
Zinc chloride ≥98.0% (by titrimetric analysis)
Zinc chloride

CAS Number: 7646-85-7
MDL Number: MFCD00011295
Molecular Formula: Cl2Zn
Molecular Weight: 136.28
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Color: White
Boiling point (°C): 732
Melting point (°C): 290

Formula: ZnCl₂
MW: 136.3 g/mol
Boiling Pt: 732 °C
Melting Pt: 290 °C
MDL Number: MFCD00011295
CAS Number: 7646-85-7
UN: 2331
ADR: 8,III
https://us.vwr.com/stibo/web/std.lang.all/74/35/28997435.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR