Diyttrium trioxide ≥97.0%
Supplier: TCI America
Synonyms:
Yttrium oxide, Yttrium (III) oxide
Y0007-100G
Y0007-25G
TCY0007-100GEA
119
USD
TCY0007-100G
TCY0007-25G
Diyttrium trioxide ≥97.0%
Diyttrium trioxide
CAS Number: 1314-36-9
MDL Number: MFCD00011473
Molecular Formula: O3Y2
Molecular Weight: 225.81
Purity/Analysis Method:
Other Product Names / Synonyms: Yttria
Formula:
Y₂O₃ |
MDL Number:
MFCD00011473 CAS Number: 1314-36-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...