4-(Trifluoromethyl)thiophenol ≥98.0%

Supplier: TCI America

Synonyms: 4-Mercaptobenzotrifluoride, 4-(Trifluoromethyl)benzene-1-thiol, 4-(Trifluoromethyl)benzenethiol

T2972-1G T2972-5G
TCT2972-1GEA 86.12 USD
TCT2972-1G TCT2972-5G
4-(Trifluoromethyl)thiophenol ≥98.0%
4-(Trifluoromethyl)thiophenol

CAS Number: 825-83-2 MDL Number: MFCD00042320 Molecular Formula: C7H5F3S Molecular Weight: 178.17 Purity/Analysis Method: 98.0% (GC,T) Form: Clear Liquid Color: Colorless Specific Gravity (20/20): 1.32


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Materials Chemistry, Self-Assembled Monolayer (SAM) Forming Agents Other Product Names / Synonyms: 4-Mercaptobenzotrifluoride, 4-(Trifluoromethyl)thiophenol

Formula: C₇H₅F₃S
Density: 1.32 (20 °C)
MDL Number: MFCD00042320
CAS Number: 825-83-2
UN: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d328dac36298f713bf4488fd89e708d500d7a0c5&vid=d328dac36298f713bf4488fd89e708d500d7a0c5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR