BAPTA-AM ≥95.0%
Supplier: TCI America
Synonyms:
Tetrakis(acetoxymethyl)-1,2-bis(2-aminophenoxy)ethane-N,N,N',N'-tetraacetate, Tetrakis(acetoxymethyl) 1,2-Bis(2-aminophenoxy)ethane-N,N,N,N-tetraacetate, 1,2-Bis(2-aminophenoxy)ethane-N,N,N',N'-tetraacetic Acid Tetrakis(acetoxymethyl) Ester, BAPTA-AM ester
CAS Number: 126150-97-8
MDL Number: MFCD00036696
Molecular Formula: C34H40N2O18
Molecular Weight: 764.69
Purity/Analysis Method:
Product Categories: Bioscience, Calcium Chelating Reagents
Other Product Names / Synonyms: 1,2-Bis(2-aminophenoxy)ethane-N,N,N',N'-tetraacetic Acid Tetrakis(acetoxymethyl) Ester, BAPTA-AM
Formula:
C₃₄H₄₀N₂O₁₈ MW: 764.69 g/mol Melting Pt: 98 °C |
MDL Number:
MFCD00036696 CAS Number: 126150-97-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...