(+/-)-Thalidomide ≥98.0%
Supplier: TCI America
Synonyms:
2-(2,6-Dioxopiperidin-3-yl)isoindoline-1,3-dione, N-(2,6-Dioxo-3-piperidinyl)phthalimide
CAS Number: 50-35-1
MDL Number: MFCD00153873
Molecular Formula: C13H10N2O4
Molecular Weight: 258.23
Purity/Analysis Method:
Product Categories: Bioscience, Immune System, Inflammation, Allergy Research Reagents, Immunosuppressant Ingredients, Antitumor Ingredients, Anti-inflammatory Ingredients
Other Product Names / Synonyms: N-(2,6-Dioxo-3-piperidinyl)phthalimide
Formula:
C₁₃H₁₀N₂O₄ Melting Pt: 276 °C |
MDL Number:
MFCD00153873 CAS Number: 50-35-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...