Tetrallyltin ≥97.0% (by GC)

Supplier: TCI America

Synonyms: Tetrallylstannane

T2009-1G T2009-5G
TCT2009-1GEA 54 USD
TCT2009-1G TCT2009-5G
Tetrallyltin ≥97.0% (by GC)
Tetrallyltin

CAS Number: 7393-43-3
Molecular Formula: C12H20Sn
Molecular Weight: 283.00
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 70
Flash Point (°C): 75
Specific Gravity (20/20): 1.19

Formula: C₁₂H₂₀Sn
MW: 283 g/mol
Boiling Pt: 70 °C
Density: 1.19 (20 °C)
Flash Pt: 75 °C
MDL Number: MFCD00008637
CAS Number: 7393-43-3
UN: 2788
ADR: 6.1,II
https://us.vwr.com/stibo/web/std.lang.all/59/50/28995950.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR