Triethylsilyl trifluoromethanesulfonate ≥98.0%

Supplier: TCI America

Synonyms: Triethylsilyl trifluoromethanesulfonate, Triethylsilyl trifluoromethanesulphonate

T1689-25G T1689-5G
TCT1689-25GEA 127.6 USD
TCT1689-25G TCT1689-5G
Triethylsilyl trifluoromethanesulfonate ≥98.0%
Triethylsilyl trifluoromethanesulphonate

CAS Number: 79271-56-0 MDL Number: MFCD00000407 Molecular Formula: C7H15F3O3SSi Molecular Weight: 264.33 Purity/Analysis Method: 98.0% (T) Form: Clear Liquid Color: Colorless Boiling point (°C): 79 Flash Point (°C): 72 Specific Gravity (20/20): 1.17


Product Categories: Synthetic Organic Chemistry, Sulfonic Acid Derivatives, Protection & Derivatization Reagents (for Synthesis), Silicon Compounds (for Synthesis) Other Product Names / Synonyms: Triethylsilyl Triflate, Trifluoromethanesulfonic Acid Triethylsilyl Ester

Formula: C₇H₁₅F₃O₃SSi
MW: 264.34 g/mol
Boiling Pt: 79 °C
Density: 1.17 (20 °C)
Flash Pt: 72 °C
MDL Number: MFCD00000407
CAS Number: 79271-56-0
UN: 3265
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=65f39215a38b6476d7bb230e135bf82e53a23aa9&vid=65f39215a38b6476d7bb230e135bf82e53a23aa9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR