Tetrabutylammonium hydroxide 40% in water

Supplier: TCI America

T1685-25G T1685-500G T1685-100G
TCT1685-25GEA 53.32 USD
TCT1685-25G TCT1685-500G TCT1685-100G
Tetrabutylammonium hydroxide 40% in water
Tetrabutylammonium hydroxide

CAS Number: 2052-49-5
MDL Number: MFCD00009425
Molecular Formula: C16H37NO
Molecular Weight: 259.48
Form: Clear Liquid
Color: Colorless
Specific Gravity (20/20): 1.00

Formula: C₁₆H₃₇NO
Density: 1.00 (20 °C)
MDL Number: MFCD00009425
CAS Number: 2052-49-5
UN: 3263
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0646a071b38d05126ef6f72baf09baa5245cf1a9&vid=f1e3ae31bc95cd81b3753242e838c18c4bf34687&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR