Tetrabutylammonium hydroxide 10% methanol
Supplier: TCI America
T0041-25ML
T0041-500ML
TCT0041-25MLEA
69
USD
TCT0041-25ML
TCT0041-500ML
Tetrabutylammonium hydroxide 10% methanol
Tetrabutylammonium hydroxide
[for non-aqueous titration]
CAS Number: 2052-49-5
MDL Number: MFCD00009425
Molecular Formula: C16H37NO
Molecular Weight: 259.48
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 11
Formula:
C₁₆H₃₇NO MW: 259.48 Melting Pt: 29.85 Density: 0.968 Flash Pt: 11 °C |
MDL Number:
MFCD00009425 CAS Number: 2052-49-5 UN: 3263 ADR: 8 II |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...