1-Decene, Standard Material for GC

Supplier: TCI
S0340-5ML
TCS0340-5MLEA 428.23 USD
TCS0340-5ML
1-Decene, Standard Material for GC
Standards Organic Standards
Form: Clear Liquid
Color: Colorless
Specific Gravity (20/20): 0.74
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR