Risperidone ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
CAS Number: 106266-06-2
MDL Number: MFCD00274576
Molecular Formula: C23H27FN4O2
Molecular Weight: 410.49
Purity/Analysis Method:
Product Categories: Synthetic Organic Chemistry, Fluorinated Bioactive Compounds, Bioscience, Inhibitors (for Pharmacological Research), Neurotransmitter Agent Ingredients, Prodrug Ingredients
Other Product Names / Synonyms: 3-[2-[4-(6-Fluoro-1,2-benzisoxazol-3-yl)-1-piperidinyl]ethyl]-6,7,8,9-tetrahydro-2-methyl-4H-pyrido[1,2-a]pyrimidin-4-one
Formula:
C₂₃H₂₇FN₄O₂ Melting Pt: 172 °C |
MDL Number:
MFCD00274576 CAS Number: 106266-06-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...