Heptafluorobutyric acid ≥99.0% (by titrimetric analysis)

Supplier: TCI America

Q0054-25G
TCQ0054-25GEA 251.96 USD
TCQ0054-25G
Heptafluorobutyric acid ≥99.0% (by titrimetric analysis)
Heptafluorobutyric acid

[Sequenation Reagent]
CAS Number: 375-22-4
MDL Number: MFCD00004171
Molecular Formula: C4HF7O2
Molecular Weight: 214.04
Purity/Analysis Method: 99.0% (T)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 120
Melting point (°C): -20
Specific Gravity (20/20): 1.66


Other Product Names / Synonyms: HFBA, Perfluorobutyric Acid

Formula: C₄HF₇O₂
MW: 214.04 g/mol
Boiling Pt: 120 °C
Melting Pt: –20 °C
Density: 1.66 (20 °C)
MDL Number: MFCD00004171
CAS Number: 375-22-4
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR