Pyrimethamine ≥98.0%
Supplier: TCI America
Synonyms:
5-(4-Chlorophenyl)-6-ethylpyrimidine-2,4-diamine, 5-(4-Chlorophenyl)-6-ethyl-2,4-pyrimidinediamine
P2037-1G
P2037-5G
TCP2037-1GEA
43
USD
TCP2037-1G
TCP2037-5G
Pyrimethamine ≥98.0%
Pyrimethamine
CAS Number: 58-14-0
MDL Number: MFCD00057350
Molecular Formula: C12H13ClN4
Molecular Weight: 248.71
Purity/Analysis Method:
Product Categories: Bioscience, Antiprotozoal Ingredients
Other Product Names / Synonyms: 5-(4-Chlorophenyl)-6-ethyl-2,4-pyrimidinediamine
Formula:
C₁₂H₁₃ClN₄ MW: 248.71 g/mol Melting Pt: 241 °C |
MDL Number:
MFCD00057350 CAS Number: 58-14-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...