Propyl-4-hydroxybenzoate ≥99.0% (by titrimetric analysis) for biochemical research

Supplier: TCI America

Synonyms: Propyl 4-hydroxybenzoate, Propyl parahydroxybenzoate, Propylparaben, 4-Hydroxybenzoic acid propyl ester

P1955-25G P1955-5G
TCP1955-25GEA 46.81 USD
TCP1955-25G TCP1955-5G
Propyl-4-hydroxybenzoate ≥99.0% (by titrimetric analysis) for biochemical research
Propyl 4-hydroxybenzoate

[for Biochemical Research]
CAS Number: 94-13-3
MDL Number: MFCD00002354
Molecular Formula: C10H12O3
Molecular Weight: 180.20
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Color: White
Melting point (°C): 98
Flash Point (°C): 185


Product Categories: Bioscience, Preservatives for Biochemical Research
Other Product Names / Synonyms: 4-Hydroxybenzoic Acid Propyl Ester, Propylparaben

Formula: C₁₀H₁₂O₃
MW: 180.2 g/mol
Boiling Pt: 292
Melting Pt: 95
Flash Pt: 185 °C
MDL Number: MFCD00002354
CAS Number: 94-13-3
Merck Index: 13,07958
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=002aa80f621e575dd3d318e9a56bd30f0a150c9b&vid=44346c1c0b9b8ef0afee97cb780f709798acb35c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR