Potassium sorbate ≥99.0% (by titrimetric analysis) for biochemical research
Supplier: TCI America
Synonyms:
(2E,4E)-2,4-Hexadienoic acid potassium salt
[for Biochemical Research]
CAS Number: 24634-61-5
MDL Number: MFCD00016546
Molecular Formula: C6H7KO2
Molecular Weight: 150.22
Purity/Analysis Method: >99.0% (T)
Form: Granul
Color: White
Product Categories: Bioscience, Preservatives for Biochemical Research
Other Product Names / Synonyms: Potassium 2,4-Hexadienoate, Sorbic Acid Potassium Salt
Formula:
CH₃CH=CHCH=CHCOOK MW: 150.22 g/mol |
MDL Number:
MFCD00016546 CAS Number: 24634-61-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...