Pinostilbene ≥97.0% (by GC)
Supplier: TCI America
Synonyms:
(E)-3-(4-Hydroxystyryl)-5-methoxyphenol
CAS Number: 42438-89-1
MDL Number: MFCD12407153
Molecular Formula: C15H14O3
Molecular Weight: 242.27
Purity/Analysis Method:
Form: Crystal
Color: White
Melting point (°C): 119
Storage Temperature: 0-10°C
Product Categories: Bioscience, Inhibitors (for Pharmacological Research), Aromatic Polyketides, Polyphenols (Phytochemicals)
Other Product Names / Synonyms: 3,4'-Dihydroxy-5-methoxy-trans-stilbene
Formula:
C₁₅H₁₄O₃ MW: 242.269 g/mol Melting Pt: 119 °C |
MDL Number:
MFCD12407153 CAS Number: 42438-89-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...