2,5-Diaminobenzenesulfonic acid ≥98.0%

Supplier: TCI America

Synonyms: 1,4-Phenylenediamine-2-sulfonic acid, 1,4-Phenylenediamine-2-sulphonic acid, 2,5-Diaminobenzenesulfonic acid, 2,5-Diaminobenzenesulphonic acid

P1691-25G P1691-5G
TCP1691-25GEA 137 USD
TCP1691-25G TCP1691-5G
2,5-Diaminobenzenesulfonic acid ≥98.0%
2,5-Diaminobenzenesulphonic acid

CAS Number: 88-45-9 MDL Number: MFCD00007904 Molecular Formula: C6H8N2O3S Molecular Weight: 188.20 Purity/Analysis Method: 98.0% (HPLC,T) Form: Crystal Color: White Melting point (°C): 300


Other Product Names / Synonyms: 2,5-Diaminobenzenesulfonic Acid

Formula: C₆H₈N₂O₃S
MW: 188.21
Boiling Pt: 500
Melting Pt: 300 °C
MDL Number: MFCD00007904
CAS Number: 88-45-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ed6bdec0502797a88b9742880b62f0d9324bba5f&vid=ed6bdec0502797a88b9742880b62f0d9324bba5f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR