Pentafluorobenzyl bromide ≥98.0% (by GC)

Supplier: TCI America

Synonyms: α-Bromo-2,3,4,5,6-pentafluorotoluene 2,3,4,5,6-Pentafluorobenzyl bromide

P0809-25G P0809-1G P0809-5G
TCP0809-5GEA 70.18 USD
TCP0809-5G TCP0809-25G TCP0809-1G
Pentafluorobenzyl bromide ≥98.0% (by GC)
Pentafluorobenzyl bromide

CAS Number: 1765-40-8
MDL Number: MFCD00000299
Molecular Formula: C7H2BrF5
Molecular Weight: 260.99
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 176
Flash Point (°C): 83
Freezing point (°C): 18
Specific Gravity (20/20): 1.85

Formula: (C₆F₅)CH₂Br
MW: 260.99 g/mol
Boiling Pt: 176 °C
Density: 1.85 (20 °C)
Flash Pt: 83 °C
MDL Number: MFCD00000299
CAS Number: 1765-40-8
UN: 3265
ADR: 8,II
https://us.vwr.com/stibo/web/std.lang.all/14/76/28991476.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR