1H-[1,2,4]Oxadiazolo[4,3-a]quinoxalin-1-one ≥95.0% (by GC)

Supplier: TCI America

Synonyms: ODQ

O0400-25MG
TCO0400-25MGEA 164.2 USD
TCO0400-25MG
1H-[1,2,4]Oxadiazolo[4,3-a]quinoxalin-1-one ≥95.0% (by GC)
1H-[1,2,4]Oxadiazolo[4,3-a]quinoxalin-1-one

CAS Number: 41443-28-1
MDL Number: MFCD00792620
Molecular Formula: C9H5N3O2
Molecular Weight: 187.16
Purity/Analysis Method: 95.0% (GC)
Form: Crystal
Melting point (°C): 162
Storage Temperature: 0-10°C


Product Categories: Bioscience, Inhibitors (for Pharmacological Research)
Other Product Names / Synonyms: ODQ

Formula: C₉H₅N₃O₂
MW: 187.16 g/mol
Melting Pt: 162 °C
MDL Number: MFCD00792620
CAS Number: 41443-28-1
https://us.vwr.com/stibo/web/std.lang.all/06/29/28990629.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR