Osmium tetraoxide 4% in water

Supplier: TCI America

Synonyms: Osmium (VIII) oxide Osmium tetraoxide Osmic Acid AnhydrideOsmium Tetroxide

O0308-10ML
TCO0308-10MLEA 157.77 USD
TCO0308-10ML
Osmium tetraoxide 4% in water
Osmium tetraoxide

CAS Number: 20816-12-0
MDL Number: MFCD00011150
Molecular Formula: O4Os
Molecular Weight: 254.23
Form: Clear Liquid
Color: Slightly Pale Yellow
Specific Gravity (20/20): 1.04
Storage Temperature: 0-10°C


Other Product Names / Synonyms: Osmium(VIII) Oxide

Formula: O₄Os
MW: 254.23 g/mol
Density: 1.04 (20 °C)
MDL Number: MFCD00011150
CAS Number: 20816-12-0
UN: 2471
ADR: 6.1,I
https://us.vwr.com/stibo/web/std.lang.all/05/71/28990571.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR