3-Methylcyclopentadecanone ≥97.0%

Supplier: TCI America

Synonyms: 3-Methylcyclopentadecanone, Muscone

M2611-1G M2611-200MG
TCM2611-1GEA 489 USD
TCM2611-1G TCM2611-200MG
3-Methylcyclopentadecanone ≥97.0%
3-Methylcyclopentadecanone

CAS Number: 541-91-3 MDL Number: MFCD00211114 Molecular Formula: C16H30O Molecular Weight: 238.42 Purity/Analysis Method: 97.0% (GC) Form: Clear Liquid Boiling point (°C): 95 Melting point (°C): -15 Specific Gravity (20/20): 0.92 Specific rotation [a]20/D: -13 deg (neat) Lambda max.: 285 nm (EtOH)


Other Product Names / Synonyms: 3-Methylcyclopentadecanone

Formula: C₁₆H₃₀O
MW: 238.41
Boiling Pt: 80
Melting Pt: -15
Density: 0.92 (20 °C)
MDL Number: MFCD00211114
CAS Number: 541-91-3
Merck Index: 12,06392
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1d2481f4d4256fee3d90f6d7db780864d2004cbc&vid=1d2481f4d4256fee3d90f6d7db780864d2004cbc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR