Manganese(II) chloride tetrahydrate 98,0
Supplier: TCI America
Synonyms:
Manganese dichloride tetrahydrate, Manganese chloride (tetrahydrate), Manganese(II) chloride tetrahydrate, Manganous chloride tetrahydrate, Manganese chloride tetrahydrate, Manganese Chloride, Tetrahydrate
M2095-25G
M2095-500G
TCM2095-25GEA
25
USD
TCM2095-25G
TCM2095-500G
Manganese(II) chloride tetrahydrate 98,0
Manganese (II) chloride tetrahydrate
CAS Number: 13446-34-9
Molecular Formula: Cl2Mn
Molecular Weight: 125.84
Purity/Analysis Method:
Formula:
MnCl₂·4H₂O MW: 197.9 g/mol Boiling Pt: 1190 Melting Pt: 650 °C |
MDL Number:
MFCD00149792 CAS Number: 13446-34-9 UN: 3077 ADR: 9 III Merck Index: 13,05751 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...