Methanol-[D₄] (99.8% D)

Supplier: TCI America
Danger

Synonyms: Carbinol-D4 Hydroxy methane-D4 Methyl alcohol-D4 Wood alcohol-D4 Methyl-D3 alcohol-D Methanol-[²H₄]

M1869-10ML M1869-10G
TCM1869-10MLEA 113.16 USD
TCM1869-10ML TCM1869-10G
Methanol-[D₄] (99.8% D)
Methanol-[D4]

CAS Number: 811-98-3
MDL Number: MFCD00044637
Molecular Formula: CD4O
Molecular Weight: 36.07
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 66
Melting point (°C): -98
Flash Point (°C): 11
Specific Gravity (20/20): 0.89


Other Product Names / Synonyms: Tetradeuteromethanol, Methyl Alcohol-d4

Formula: D₃COD
MW: 36.01 g/mol
Boiling Pt: 66 °C
Melting Pt: −98 °C
Density: 0.89 (20 °C)
Flash Pt: 11 °C
MDL Number: MFCD00044637
CAS Number: 811-98-3
EINECS: 212-378-6
UN: 1230
ADR: 3,II

Order Now

Specification Test Results

Appearance Colourless clear liquid
Degree of deuteration min. 99.8 atom%D

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR