3-Methylacetylacetone ≥96.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 3-Methyl-2,4-pentanedione

M1132-5ML M1132-25ML
TCM1132-25MLEA 584.64 USD
TCM1132-25ML TCM1132-5ML
3-Methylacetylacetone ≥96.0% (by titrimetric analysis)
3-Methylacetylacetone

CAS Number: 815-57-6
MDL Number: MFCD00008762
Molecular Formula: C6H10O2
Molecular Weight: 114.14
Purity/Analysis Method: 96.0% (T)
Form: Clear Liquid
Boiling point (°C): 172
Flash Point (°C): 56
Specific Gravity (20/20): 0.98
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Oxidation, Ligands
Other Product Names / Synonyms: 1,1-Diacetylethane, Methylacetylacetone

Formula: C₆H₁₀O₂
MW: 114.14 g/mol
Boiling Pt: 172 °C
Density: 0.98 (20 °C)
Flash Pt: 56 °C
MDL Number: MFCD00008762
CAS Number: 815-57-6
UN: 1224
ADR: 3,III
https://us.vwr.com/stibo/web/std.lang.all/48/30/28974830.jpg

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR