Methylene dithiocyanate ≥99.0% (by GC)

Supplier: TCI America

Synonyms: Methylene BisthiocyanateDithiocyanatomethane

M1020-25G
TCM1020-25GEA 146.11 USD
TCM1020-25G
Methylene dithiocyanate ≥99.0% (by GC)
Methylene dithiocyanate

CAS Number: 6317-18-6
MDL Number: MFCD00001833
Molecular Formula: C3H2N2S2
Molecular Weight: 130.18
Purity/Analysis Method: 99.0% (GC)
Form: Crystal
Melting point (°C): 105


Other Product Names / Synonyms: Dithiocyanatomethane, Methylene Bisthiocyanate

Formula: C₃H₂N₂S₂
MW: 130.18 g/mol
Melting Pt: 105 °C
MDL Number: MFCD00001833
CAS Number: 6317-18-6
https://us.vwr.com/stibo/web/std.lang.all/46/36/28974636.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR