Methyl orange ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: Helianthine

M0489-25G
TCM0489-25GEA 107.05 USD
TCM0489-25G
Methyl orange ≥98.0% (by HPLC)
Methyl orange

CAS Number: 547-58-0
MDL Number: MFCD00007502
Molecular Formula: C14H15N3O3S
Molecular Weight: 327.33
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal
Lambda max.: 465 nm (pH 4.8 buffer sol.)


Product Categories: Analytical Science, Indicators (pH)
Other Product Names / Synonyms: Orange III

Formula: C₁₄H₁₄N₃NaO₃S
MW: 327.34 g/mol
MDL Number: MFCD00007502
CAS Number: 547-58-0
UN: 3143
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/39/52/28973952.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR