Maleimide ≥98.0%

Supplier: TCI America

Synonyms: 1H-Pyrrole-2,5-dione

M0018-25G M0018-5G
TCM0018-25GEA 191 USD
TCM0018-25G TCM0018-5G
Maleimide ≥98.0%
Maleimide

CAS Number: 541-59-3 MDL Number: MFCD00005494 Molecular Formula: C4H3NO2 Molecular Weight: 97.07 Purity/Analysis Method: 98.0% (T) Form: Crystal Boiling point (°C): 125 Melting point (°C): 94 Lambda max.: 280 nm (EtOH)

Formula: C₄H₃NO₂
Boiling Pt: 125 °C
Melting Pt: 94 °C
MDL Number: MFCD00005494
CAS Number: 541-59-3
UN: 2923
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d318089a16121dcd366c3a85e93bdbeedfff951b&vid=d318089a16121dcd366c3a85e93bdbeedfff951b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR