(1R,2S,5R)-2-Isopropyl-5-methylcyclohexyl-(S)-2-hydroxypropionate 98,0

Supplier: TCI America

Synonyms: (S)-2-Hydroxypropionic acid (1R,2S,5R)-2-Isopropyl-5-methylcyclohexyl Ester, (S)-(1R,2S,5R)-2-Isopropyl-5-methylcyclohexyl 2-hydroxypropanoate, L-Menthyl L-Lactate

I0889-25G I0889-5G
TCI0889-25GEA 108.8 USD
TCI0889-25G TCI0889-5G
(1R,2S,5R)-2-Isopropyl-5-methylcyclohexyl-(S)-2-hydroxypropionate 98,0
(1R,2S,5R)-2-Isopropyl-5-methylcyclohexyl-(S)-2-hydroxypropionate

CAS Number: 61597-98-6 Molecular Formula: C13H24O3 Molecular Weight: 228.33 Purity/Analysis Method: 98.0% (GC) Form: Crystal Melting point (°C): 45 Specific rotation [a]20/D: -82 deg (C=5, MeOH) Lambda max.: 233 nm (CH2Cl2)


Product Categories: Bioscience, Monocyclic Monoterpenes Other Product Names / Synonyms: (S)-2-Hydroxypropionic Acid (1R,2S,5R)-2-Isopropyl-5-methylcyclohexyl Ester, L-Menthyl L-Lactate

Formula: C₁₃H₂₄O₃
MW: 228.33 g/mol
Melting Pt: 45 °C
MDL Number: MFCD27977194
CAS Number: 61597-98-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=30fa5f82c4abf1243267aed6d87c2c1aae931a9a&vid=30fa5f82c4abf1243267aed6d87c2c1aae931a9a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR