(S)-(-)-1-Phenylethyl isocyanate ≥98.0%

Supplier: TCI America

Synonyms: (S)-(-)-α-Phenethyl isocyanate, (S)-(-)-α-Methylbenzyl isocyanate, (S)-1-phenylethylisocyanate, (S)-(-)-1-Phenethyl isocyanate

I0335-1G I0335-5G
TCI0335-1GEA 50 USD
TCI0335-1G TCI0335-5G
(S)-(-)-1-Phenylethyl isocyanate ≥98.0%
(S)-(-)-1-Phenylethyl isocyanate

CAS Number: 14649-03-7 MDL Number: MFCD00002036 Molecular Formula: C9H9NO Molecular Weight: 147.18 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Boiling point (°C): 98 Flash Point (°C): 65 Specific Gravity (20/20): 1.05 Specific rotation [a]20/D: -10 deg (neat) Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Optical Resolving Agents, Optical Resolving Agents, Chiral Building Blocks, Analytical Science, e.e. Determination (HPLC Labeling Reagents), e.e. / Absolute Configuration Determination (NMR) Other Product Names / Synonyms: Isocyanic Acid (S)-(-)-alpha-Methylbenzyl Ester, Isocyanic Acid (S)-(-)-1-Phenylethyl Ester, (S)-(-)-1-Phenylethyl Isocyanate

Formula: C₉H₉NO
Boiling Pt: 98 °C
Density: 1.05 (20 °C)
Flash Pt: 65 °C
MDL Number: MFCD00002036
CAS Number: 14649-03-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=284ef7530654509fc19340bf6c20d584c42aabb8&vid=284ef7530654509fc19340bf6c20d584c42aabb8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR