bisBenzimide H 33258 trihydrochloride hydrate (Hoechst 33258) ≥98.0% (by HPLC) fluorescent dye, for biochemical research

Supplier: TCI America

Synonyms: Bisbenzimide H 33258

H1343-25MG
TCH1343-25MGEA 81.5 USD
TCH1343-25MG
bisBenzimide H 33258 trihydrochloride hydrate (Hoechst 33258) ≥98.0% (by HPLC) fluorescent dye, for biochemical research
bisBenzimide H 33258 trihydrochloride hydrate (Hoechst 33258)
Formula: C₂₅H₂₄N₆O.3HCl.xH₂O
MW: 533.88 g/mol (anhy)
Melting Pt: 280 °C (lit.)
Storage Temperature: Freezer
MDL Number: MFCD00012679
CAS Number: 23491-45-4
https://us.vwr.com/stibo/web/std.lang.all/13/45/28971345.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR