N-Hydroxysulfosuccinimide sodium salt (Sulfo-NHS) ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
N-Hydroxysulfosuccinimide sodium salt
, sulfo-N-Hydroxysuccinimide
, Hydroxy-2,5-dioxopyrrolidine-3-sulphonic acid sodium salt
, N-Hydroxy sulfosuccinimide sodium salt
, Sulfo NHS
, N-Hydroxysulphosuccinimide sodium salt
, Hydroxy-2,5-dioxopyrrolidine-3-sulfonic acid sodium salt
, N-Hydroxy sulphosuccinimide sodium salt
, Sulpho-N-Hydroxysuccinimide
CAS Number: 106627-54-7
MDL Number: MFCD00043100
Molecular Formula: C4H5NO6S
Molecular Weight: 217.13
Purity/Analysis Method:
Form: Crystal
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Condensation & Active Esterification, Bioscience, Crosslinkers
Other Product Names / Synonyms: Sulfo-NHS
Formula:
C4H4NNaO6S MW: 217.13 g/mol |
MDL Number:
MFCD00043100 CAS Number: 106627-54-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...