Hydrogen chloride ∼1 mol/L in ethyl ether
Supplier: TCI America
H1061-500ML
H1061-25ML
TCH1061-25MLEA
39.94
USD
TCH1061-25ML
TCH1061-500ML
Hydrogen chloride ∼1 mol/L in ethyl ether
Hydrogen chloride
CAS Number: 7647-01-0
MDL Number: MFCD00011324
Molecular Formula: HCl
Molecular Weight: 36.46
Form: Clear Liquid
Color: Colorless
Flash Point (°C): -40
Specific Gravity (20/20): 0.73
Storage Temperature: 0-10°C
Formula:
HCl MW: 36.46 g/mol Density: 0.73 (20 °C) Flash Pt: –40 °C |
MDL Number:
MFCD00011324 CAS Number: 7647-01-0 UN: 1050 ADR: 2, |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...