Gemfibrozil ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
2,2-Dimethyl-5-(2,5-dimethylphenoxy)pentanoic acid
CAS Number: 25812-30-0
MDL Number: MFCD00079335
Molecular Formula: C15H22O3
Molecular Weight: 250.34
Purity/Analysis Method:
Form: Crystal
Boiling point (°C): 159
Melting point (°C): 61
Product Categories: Bioscience, Antilipemic Ingredients
Other Product Names / Synonyms: 2,2-Dimethyl-5-(2,5-dimethylphenoxy)pentanoic Acid, 2,2-Dimethyl-5-(2,5-dimethylphenoxy)valeric Acid
Formula:
C₁₅H₂₂O₃ MW: 250.34 g/mol Boiling Pt: 159 °C Melting Pt: 61 °C |
MDL Number:
MFCD00079335 CAS Number: 25812-30-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...