4-(4-Fluorophenyl)-6-isopropyl-2-(N-methyl-N-methanesulfonylamino)-5-pyrimidinecarboxaldehyde ≥98.0%

Supplier: TCI America

Synonyms: N-[4-(4-Fluorophenyl)-5-formyl-6-isopropyl-2-pyrimidinyl]-N-methylmethanesulphonamide, N-[4-(4-Fluorophenyl)-5-formyl-6-isopropyl-2-pyrimidinyl]-N-methylmethanesulfonamide, N-[4-(4-Fluorophenyl)-5-formyl-6-(1-methylethyl)-2-pyrimidinyl]-N-methyl-methanesulphonamide, N-[4-(4-Fluorophenyl)-5-formyl-6-(1-methylethyl)-2-pyrimidinyl]-N-methyl-methanesulfonamide, N-(4-(4-Fluorophenyl)-5-formyl-6-isopropylpyrimidin-2-yl)-N-methylmethanesulphonamide, N-(4-(4-Fluorophenyl)-5-formyl-6-isopropylpyrimidin-2-yl)-N-methylmethanesulfonamide

F0867-25G F0867-5G
TCF0867-25GEA 86 USD
TCF0867-25G TCF0867-5G
4-(4-Fluorophenyl)-6-isopropyl-2-(N-methyl-N-methanesulfonylamino)-5-pyrimidinecarboxaldehyde ≥98.0%
4-(4-Fluorophenyl)-6-isopropyl-2-(N-methyl-N-methanesulfonylamino)-5-pyrimidinecarboxaldehyde

CAS Number: 147118-37-4 MDL Number: MFCD08458342 Molecular Formula: C16H18FN3O3S Molecular Weight: 351.40 Purity/Analysis Method: 98.0% (HPLC,N) Form: Crystal Melting point (°C): 180

Formula: C₁₆H₁₈FN₃O₃S
MW: 351.40 g/mol
Melting Pt: 180 °C
MDL Number: MFCD08458342
CAS Number: 147118-37-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aad37155c192208d6778a52f6bc46e9db5138173&vid=aad37155c192208d6778a52f6bc46e9db5138173&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR