Emtricitabine ≥98.0%

Supplier: TCI America

Synonyms: 4-Amino-5-fluoro-1-((2R,5S)-2-(hydroxymethyl)-1,3-oxathiolan-5-yl)pyrimidin-2(1H)-one

E1007-250MG E1007-25MG
TCE1007-250MGEA 309 USD
TCE1007-250MG TCE1007-25MG
Emtricitabine ≥98.0%
Emtricitabine

CAS Number: 143491-57-0 MDL Number: MFCD00870151 Molecular Formula: C8H10FN3O3S Molecular Weight: 247.24 Purity/Analysis Method: 98.0% (HPLC,T) Form: Crystal Melting point (°C): 155 Specific rotation [a]20/D: -140 deg (C=0.25, MeOH) Lambda max.: 280 nm (H2O)

Formula: C₈H₁₀FN₃O₃S
MW: 247.24 g/mol
Melting Pt: 155 °C
MDL Number: MFCD00870151
CAS Number: 143491-57-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6810134e1e9fc35d7639e3edeae397dda53cb3ab&vid=6810134e1e9fc35d7639e3edeae397dda53cb3ab&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR