4(5)-Ethylimidazole ≥98.0% (by GC, titration analysis)
Supplier: TCI America
E0984-1G
E0984-5G
TCE0984-1GEA
145.4
USD
TCE0984-1G
TCE0984-5G
4(5)-Ethylimidazole ≥98.0% (by GC, titration analysis)
4(5)-Ethylimidazole
CAS Number: 19141-85-6
Molecular Formula: C5H8N2
Molecular Weight: 96.13
Purity/Analysis Method:
Formula:
C₅H₈N₂ |
MDL Number:
MFCD20617036 CAS Number: 19141-85-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...