Ethylene glycolbis(propionitrile)ether ≥97.0%
Supplier: TCI America
Synonyms:
Ethylene Glycol Bis(propionitrile) Ether, 3,3'-(Ethylenedioxy)dipropionitrile, Ethylene Glycol Bis(2-cyanoethyl) Ether, 1,2-Bis(2-cyanoethoxy)ethane, Ethyleneglycolbis(propionitrile)ether
CAS Number: 3386-87-6
MDL Number: MFCD00019871
Molecular Formula: C8H12N2O2
Molecular Weight: 168.20
Purity/Analysis Method:
Product Categories: Materials Chemistry, Secondary Battery Research Reagents
Other Product Names / Synonyms: 1,2-Bis(2-cyanoethoxy)ethane, 3,3'-(Ethylenedioxy)dipropionitrile, Ethylene Glycol Bis(2-cyanoethyl) Ether
Formula:
C₈H₁₂N₂O₂ Boiling Pt: 165 °C Density: 1.07 (20 °C) |
MDL Number:
MFCD00019871 CAS Number: 3386-87-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...