Dess-Martin periodinane 8 - 12% in dichloromethane

Supplier: TCI America

Synonyms: 1,1-Dihydro-1,1,1-triacetoxy-1,2-benziodoxol-3(1H)-one, DMP, (1,1,1-Triacetoxy)-1,1-dihydro-1,2-benziodoxol-3(1H)-one, Acetic acid 1,1-diacetoxy-3-oxo-1l5-ioda-2-oxa-indan-1-yl ester

D4477-250ML D4477-25ML
TCD4477-250MLEA 628.7 USD
TCD4477-250ML TCD4477-25ML
Dess-Martin periodinane 8 - 12% in dichloromethane
Dess-Martin periodinane

CAS Number: 87413-09-0
MDL Number: MFCD00130127
Molecular Formula: C13H13IO8
Molecular Weight: 424.14
Form: Clear Liquid
Color: Colorless
Flash Point (°C): -11
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 1,1,1-Triacetoxy-1,1-dihydro-1,2-benziodoxol-3(1H)-one

Formula: C₁₃H₁₃IO₈
Flash Pt: –11 °C
MDL Number: MFCD00130127
CAS Number: 87413-09-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aa18620f1b3b69de396787de83bada1577ceaf4e&vid=8993fe4100bf99a12786b3d71eb33028b9061f57&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR