2,2-Diphenyl-1-picrylhydrazyl (free radical) ≥97.0%

Supplier: TCI America
Danger

Synonyms: 2,2-Diphenyl-1-(2,4,6-trinitrophenyl)hydrazyl free radical, DPPH free radical

D4313-1G D4313-5G
TCD4313-1GEA 127.57 USD
TCD4313-1G TCD4313-5G
2,2-Diphenyl-1-picrylhydrazyl (free radical) ≥97.0%
2,2-Diphenyl-1-picrylhydrazyl (free radical)

Form: Crystal
Melting point (°C): 139
Lambda max.: 515 nm (MeOH)
Storage Temperature: 0-10°C


Product Categories: Materials Chemistry, Organic Radicals (Magnetic Materials), Polymerization Inhibitors, Bioscience, ROS/RNS Detection Reagents, Redox Research Reagents Other Product Names / Synonyms: DPPH

Formula: C₁₈H₁₂N₅O₆
Melting Pt: 139 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007231
CAS Number: 1898-66-4
EINECS: 217-591-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5181128be0ae2910b83952379e8bafa1b60bce30&vid=5181128be0ae2910b83952379e8bafa1b60bce30&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR